accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OH3CBr

CAS RN 40180-80-1

IUPAC Name: 1-(3-bromo-4-methylphenyl)ethan-1-one
Synonyms: 1-(3-bromo-4-methylphenyl)ethanone
Molecular Weight (g/mol): 213.07
Molecular Formula: C9H9BrO
InChi Key: PDJLFESXPVLVMR-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Br)=C(C)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)