accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40186-71-8

IUPAC Name: 2,2',3,3',4,5',6,6'-octachloro-1,1'-biphenyl
Synonyms: 2,2',3,3',4,5',6,6'-octachloro-1,1'-biphenyl
Molecular Weight (g/mol): 429.75
Molecular Formula: C12H2Cl8
InChi Key: LJQOBQLZTUSEJA-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C(Cl)=C1Cl)C1=C(Cl)C(Cl)=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results