CAS RN 40188-45-2
CAS RN 40188-45-2
IUPAC Name:
N-(3-acetyl-4-hydroxyphenyl)butanamide
Synonyms:
N-(3-acetyl-4-hydroxyphenyl)butanamide
Molecular Weight (g/mol):
221.26
Molecular Formula:
C12H15NO3
InChi Key:
FGWZEOPEZISTTR-UHFFFAOYSA-N
SMILES:
CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O
1
–
1
of
1
results
1
Thermo Scientific Chemicals N-(3-Acetyl-4-hydroxyphenyl)butyramide, 97%
CAS: 40188-45-2 Molecular Formula: C12H15NO3 Molecular Weight (g/mol): 221.26 MDL Number: MFCD00798556 InChI Key: FGWZEOPEZISTTR-UHFFFAOYSA-N Synonym: 2-acetyl-4-butyramidophenol,n-3-acetyl-4-hydroxyphenyl butanamide,n-3-acetyl-4-hydroxyphenyl butyramide,ac-hophe-oh,unii-14qjb65r4v,3'-acetyl-4'-hydroxybutyranilide,butanamide, n-3-acetyl-4-hydroxyphenyl,5'-butyramido-2'-hydroxyacetophenone,acebutolol impurity c,n-3-acetyl-4-hydroxyphenyl butanamid PubChem CID: 736331 IUPAC Name: N-(3-acetyl-4-hydroxyphenyl)butanamide SMILES: CCCC(=O)NC1=CC=C(O)C(=C1)C(C)=O