accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFF

CAS RN 402-45-9

IUPAC Name: 4-(trifluoromethyl)phenol
Synonyms: α,α,α-trifluoro-P-cresol P-trifluoromethylphenol α,α,α-trifluoro-p-cresol
Molecular Weight (g/mol): 162.11
Molecular Formula: C7H5F3O
InChi Key: BAYGVMXZJBFEMB-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)