accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 402-52-8

H3COFFF

CAS RN 402-52-8

IUPAC Name: 1-methoxy-4-(trifluoromethyl)benzene
Synonyms: 1-methoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 176.14
Molecular Formula: C8H7F3O
InChi Key: CFIPQRIPCRRISV-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results