accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOO

CAS RN 4021-07-2

IUPAC Name: 3-methylpyridine-2-carboxylic acid
Synonyms: 3-methylpyridine-2-carboxylic acid
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: LMHIBYREWJHKNZ-UHFFFAOYSA-N
SMILES: CC1=CC=CN=C1C(O)=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)