accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4021-50-5

IUPAC Name: 4-[(trifluoromethyl)sulfanyl]benzaldehyde
Synonyms: 4-[(trifluoromethyl)sulfanyl]benzaldehyde
Molecular Weight (g/mol): 206.18
Molecular Formula: C8H5F3OS
InChi Key: OPAXHVNZGUGCKW-UHFFFAOYSA-N
SMILES: FC(F)(F)SC1=CC=C(C=O)C=C1
Boiling Point 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)