accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40210-84-2

IUPAC Name: 2-(2H-1,3-dithiol-2-ylidene)-2H-1,3-dithiole; 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
Synonyms: tetracyanoquinodimethane; tetrathiafulvalene
Molecular Weight (g/mol): 408.53
Molecular Formula: C18H8N4S4
InChi Key: OIXMVDHMELKBDX-UHFFFAOYSA-N
SMILES: S1C=CSC1=C1SC=CS1.N#CC(C#N)=C1C=CC(C=C1)=C(C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results