accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4023-65-8

IUPAC Name: (1E)-prop-1-ene-1,2,3-tricarboxylic acid
Synonyms: trans-aconitic acid
Molecular Weight (g/mol): 174.11
Molecular Formula: C6H6O6
InChi Key: GTZCVFVGUGFEME-HNQUOIGGSA-N
SMILES: OC(=O)C\C(=C/C(O)=O)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)