accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHOO

CAS RN 40248-63-3

IUPAC Name: 2-{[5-methyl-2-(propan-2-yl)cyclohexyl]oxy}acetic acid
Synonyms: [(2-isopropyl-5-methylcyclohexyl)oxy]acetic acid
Molecular Weight (g/mol): 214.31
Molecular Formula: C12H22O3
InChi Key: CILPHQCEVYJUDN-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results