accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOSClOO

CAS RN 4025-64-3

IUPAC Name: 3-(chlorosulfonyl)benzoic acid
Synonyms: 3-(chlorosulfonyl)benzoic acid
Molecular Weight (g/mol): 220.62
Molecular Formula: C7H5ClO4S
InChi Key: LMRKXSDOAFUINK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)