CAS RN 4027-54-7
IUPAC Name:
3-phenyl-5-(trifluoromethyl)-1H-pyrazole
Synonyms:
3-phenyl-5-(trifluoromethyl)-1H-pyrazole
Molecular Weight (g/mol):
212.18
Molecular Formula:
C10H7F3N2
InChi Key:
KJKWJOAARUHGNU-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(=NN1)C1=CC=CC=C1
1
–
1
of
1
results
1
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole 98.0+%, TCI America™
CAS: 4027-54-7 Molecular Formula: C10H7F3N2 Molecular Weight (g/mol): 212.18 MDL Number: MFCD00156064 InChI Key: KJKWJOAARUHGNU-UHFFFAOYSA-N PubChem CID: 611322 IUPAC Name: 3-phenyl-5-(trifluoromethyl)-1H-pyrazole SMILES: FC(F)(F)C1=CC(=NN1)C1=CC=CC=C1