accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SNOO

CAS RN 40283-41-8

IUPAC Name: 2-amino-1,3-thiazole-4-carboxylate
Synonyms: 2-amino-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 143.14
Molecular Formula: C4H3N2O2S
InChi Key: FCLDUALXSYSMFB-UHFFFAOYSA-M
SMILES: NC1=NC(=CS1)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)