accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3C

CAS RN 40291-39-2

IUPAC Name: ethyl 2-(2-methylphenyl)acetate
Synonyms: ethyl 2-(2-methylphenyl)acetate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: HTDQOYWJDUMNHX-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=CC=C1C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)