accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 402960-38-7

IUPAC Name: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
Synonyms: 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
Molecular Weight (g/mol): 221.07
Molecular Formula: C10H16BN3O2
InChi Key: BPQVMIDUTRJYSC-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(N)N=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)