accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOO

CAS RN 403-32-7

IUPAC Name: 2-(4-fluorophenyl)-2-oxoacetaldehyde
Synonyms: 2-(4-fluorophenyl)-2-oxoacetaldehyde
Molecular Weight (g/mol): 152.12
Molecular Formula: C8H5FO2
InChi Key: IPWSCROFORAGJW-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results