accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON

CAS RN 4030-18-6

IUPAC Name: 1-(pyrrolidin-1-yl)ethan-1-one
Synonyms: 1-(pyrrolidin-1-yl)ethanone
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: LNWWQYYLZVZXKS-UHFFFAOYSA-N
SMILES: CC(=O)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results