accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40307-11-7

IUPAC Name: 1-ethyl-4-ethynylbenzene
Synonyms: 1-ethyl-4-ethynylbenzene
Molecular Weight (g/mol): 130.19
Molecular Formula: C10H10
InChi Key: ZNTJVJSUNSUMPP-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C#C
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)