accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHOHO

CAS RN 40348-66-1

IUPAC Name: butane-1,2-diol
Synonyms: 1,2-butanediol α-butylene glycol α-butyleneglycol 1,2-butylene glycol ethylethylene glycol 1,2-(dihydroxy)butane
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: BMRWNKZVCUKKSR-UHFFFAOYNA-N
SMILES: CCC(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results