accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 403483-19-2

IUPAC Name: 33-(3-dodecylphenyl)-32-methyl-32-azadotriacontacyclo[9.8.42.1²,⁵.1⁶,⁵³.0³,²².0⁴,⁵².0⁷,⁵⁸.0⁸,⁶¹.0¹⁰,¹⁹.0¹²,³⁸.0¹³,¹⁸.0¹⁴,³⁷.0¹⁵,²⁸.0¹⁶,²⁶.0¹⁷,²⁰.0²¹,²⁵.0²³,⁵¹.0²⁴,⁴⁸.0²⁷,⁴⁷.0²⁹,³⁶.0³⁰,³⁴.0³⁰,⁴⁶.0³⁴,⁴³.0³⁵,⁴⁰.0³⁹,⁶⁰.0⁴¹,⁵⁹.0⁴²,⁵⁶.0⁴⁴,⁵⁵.0⁴⁵,⁴⁹.0⁵⁰,⁵⁴.0⁹,⁶³.0⁵⁷,⁶²]trihexaconta-1(19),2(63),3,5,7(58),8,10,12,14(37),15(28),16(26),17,20,22,24,27(47),29(36),35(40),38,41(59),42,44(55),45,48,50(54),51,53(62),56,60-nonacosaene
Synonyms: 33-(3-dodecylphenyl)-32-methyl-32-azadotriacontacyclo[9.8.42.1²,⁵.1⁶,⁵³.0³,²².0⁴,⁵².0⁷,⁵⁸.0⁸,⁶¹.0¹⁰,¹⁹.0¹²,³⁸.0¹³,¹⁸.0¹⁴,³⁷.0¹⁵,²⁸.0¹⁶,²⁶.0¹⁷,²⁰.0²¹,²⁵.0²³,⁵¹.0²⁴,⁴⁸.0²⁷,⁴⁷.0²⁹,³⁶.0³⁰,³⁴.0³⁰,⁴⁶.0³⁴,⁴³.0³⁵,⁴⁰.0³⁹,⁶⁰.0⁴¹,⁵⁹.0⁴²,⁵⁶.0⁴⁴,⁵⁵.0⁴⁵,⁴⁹.0⁵⁰,⁵⁴.0⁹,⁶³.0⁵⁷,⁶²]trihexaconta-1(19),2(63),3,5,7(58),8,10,12,14(37),15(28),16(26),17,20,22,24,27(47),29(36),35(40),38,41(59),42,44(55),45,48,50(54),51,53(62),56,60-nonacosaene
Molecular Weight (g/mol): 1022.18
Molecular Formula: C81H35N
InChi Key: YDAHEEMFHYNKKQ-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCC1=CC=CC(=C1)C1N(C)CC23C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%14=C%15C%16=C%17C%18=C%19C%20=C(C5=C5C%21=C6C6=C%22C%23=C(C%12=C%12C%14=C%16C%14=C%18C(C(C%14=C%23%12)=C%21%22)=C%205)C%10=C86)C12C1=C%19C%17=C2C%15=C5C%13=C%11C6=C9C(=C47)C4=C6C5=C2C1=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results