accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 403519-64-2

IUPAC Name: 2,2-bis(3-methylbutyl)propane-1,3-diol
Synonyms: 2,2-bis(3-methylbutyl)propane-1,3-diol
Molecular Weight (g/mol): 216.37
Molecular Formula: C13H28O2
InChi Key: CRJRLBBFSRNKLY-UHFFFAOYSA-N
SMILES: CC(C)CCC(CO)(CO)CCC(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results