accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40357-96-8

IUPAC Name: 5-nitrothiophene-3-carboxylate
Synonyms: 5-nitrothiophene-3-carboxylate
Molecular Weight (g/mol): 172.13
Molecular Formula: C5H2NO4S
InChi Key: VNJUNWUOAKEIKG-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CSC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results