accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40381-90-6

IUPAC Name: 2,6-dichloropyridine-3-carbonitrile
Synonyms: 2,6-dichloropyridine-3-carbonitrile
Molecular Weight (g/mol): 173
Molecular Formula: C6H2Cl2N2
InChi Key: LFCADBSUDWERJT-UHFFFAOYSA-N
SMILES: ClC1=NC(Cl)=C(C=C1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)