accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4039-82-1

IUPAC Name: [(prop-2-yn-1-yloxy)methyl]benzene
Synonyms: [(prop-2-yn-1-yloxy)methyl]benzene
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: PAQVEXAFKDWGOT-UHFFFAOYSA-N
SMILES: C#CCOCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)