accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOCH3H3COOH3C

CAS RN 40420-22-2

IUPAC Name: 1,5-diethyl 2,2-dimethyl-3-oxopentanedioate
Synonyms: 1,5-diethyl 2,2-dimethyl-3-oxopentanedioate
Molecular Weight (g/mol): 230.26
Molecular Formula: C11H18O5
InChi Key: KAMXUPAAAGYEDK-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C(C)(C)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results