accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO(S)NH

CAS RN 4043-88-3

IUPAC Name: (2S)-2,5-dihydro-1H-pyrrole-2-carboxylic acid
Synonyms: (2S)-2,5-dihydro-1H-pyrrole-2-carboxylic acid
Molecular Weight (g/mol): 113.12
Molecular Formula: C5H7NO2
InChi Key: OMGHIGVFLOPEHJ-BYPYZUCNSA-N
SMILES: OC(=O)[C@H]1NCC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results