accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4043-96-3

NOOOPOOONOONa

CAS RN 4043-96-3

IUPAC Name: sodium bis(4-nitrophenyl) phosphate
Synonyms: sodium bis-p-nitrophenyl phosphate
Molecular Weight (g/mol): 362.17
Molecular Formula: C12H8N2NaO8P
InChi Key: DELHRHCJGSTQNU-UHFFFAOYSA-M
SMILES: [Na+].[O-][N+](=O)C1=CC=C(OP([O-])(=O)OC2=CC=C(C=C2)[N+]([O-])=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results