accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHNBr

CAS RN 40432-84-6

IUPAC Name: 2-(5-bromo-1H-indol-3-yl)acetate
Synonyms: 2-(5-bromo-1H-indol-3-yl)acetate
Molecular Weight (g/mol): 253.08
Molecular Formula: C10H7BrNO2
InChi Key: WTFGHMZUJMRWBK-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CNC2=CC=C(Br)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results