accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4045-24-3

IUPAC Name: 4-methoxypiperidine
Synonyms: 4-methoxypiperidine
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: ZEYSHALLPAKUHG-UHFFFAOYSA-N
SMILES: COC1CCNCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results