accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4046-02-0

IUPAC Name: ethyl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Synonyms: ethyl ferulate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: ATJVZXXHKSYELS-FNORWQNLSA-N
SMILES: CCOC(=O)\C=C\C1=CC=C(O)C(OC)=C1
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)