accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3(E)(E)

CAS RN 40470-68-6

IUPAC Name: 4,4'-bis[(1E)-2-(2-methoxyphenyl)ethenyl]-1,1'-biphenyl
Synonyms: 4,4'-bis[(1E)-2-(2-methoxyphenyl)ethenyl]-1,1'-biphenyl
Molecular Weight (g/mol): 418.54
Molecular Formula: C30H26O2
InChi Key: HZAWHDJKNZWAAR-YHARCJFQSA-N
SMILES: COC1=CC=CC=C1\C=C\C1=CC=C(C=C1)C1=CC=C(\C=C\C2=CC=CC=C2OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results