accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40480-10-2

IUPAC Name: heptadecanoyl chloride
Synonyms: heptadecanoyl chloride
Molecular Weight (g/mol): 288.9
Molecular Formula: C17H33ClO
InChi Key: ICDQUAGMQCUEMY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC(Cl)=O
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results