accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNHNBr

CAS RN 404827-77-6

IUPAC Name: 6-bromo-1H-indazol-3-amine
Synonyms: 6-bromo-1H-indazol-3-amine
Molecular Weight (g/mol): 212.05
Molecular Formula: C7H6BrN3
InChi Key: WLDHNAMVDBASAW-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC(Br)=CC=C12
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)