accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40487-42-1

IUPAC Name: 3,4-dimethyl-2,6-dinitro-N-(pentan-3-yl)aniline
Synonyms: way up pendimethalin
Molecular Weight (g/mol): 281.31
Molecular Formula: C13H19N3O4
InChi Key: CHIFOSRWCNZCFN-UHFFFAOYSA-N
SMILES: CCC(CC)NC1=C(C=C(C)C(C)=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results