accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2CH2

CAS RN 4049-81-4

IUPAC Name: 2-methylhexa-1,5-diene
Synonyms: 2-methyl-1,5-hexadiene
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: SLQMKNPIYMOEGB-UHFFFAOYSA-N
SMILES: CC(=C)CCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results