accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHF

CAS RN 405-09-4

IUPAC Name: (2-fluoro-4-methoxyphenyl)methanol
Synonyms: (2-fluoro-4-methoxyphenyl)methanol
Molecular Weight (g/mol): 156.16
Molecular Formula: C8H9FO2
InChi Key: AZYGOIQKPGPBTM-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(CO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results