accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 405-31-2

IUPAC Name: 1-fluoro-4-[(4-fluorophenyl)disulfanyl]benzene
Synonyms: 1-fluoro-4-[(4-fluorophenyl)disulfanyl]benzene
Molecular Weight (g/mol): 254.31
Molecular Formula: C12H8F2S2
InChi Key: SLXZJIKDNHDPKL-UHFFFAOYSA-N
SMILES: FC1=CC=C(SSC2=CC=C(F)C=C2)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results