accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 405165-14-2

IUPAC Name: 4,4,5,5-tetramethyl-2-(3-octylthiophen-2-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(3-octylthiophen-2-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 322.31
Molecular Formula: C18H31BO2S
InChi Key: QGVDWQHOZUDLGM-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=C(SC=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results