accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4052-92-0

IUPAC Name: 3-(chlorosulfonyl)benzoyl chloride
Synonyms: 3-(chlorosulfonyl)benzoyl chloride
Molecular Weight (g/mol): 239.07
Molecular Formula: C7H4Cl2O3S
InChi Key: XWEBTVZIZWEJOO-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)