accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHOH3COH

CAS RN 40547-58-8

IUPAC Name: N-(4-acetyl-3-hydroxyphenyl)acetamide
Synonyms: N-(4-acetyl-3-hydroxyphenyl)acetamide
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: ZSSHNMZQDWSUJJ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(O)=C(C=C1)C(C)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results