accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3COBOHHO

CAS RN 405520-68-5

IUPAC Name: [4-(dimethylcarbamoyl)phenyl]boronic acid
Synonyms: 4-(dimethylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 193.01
Molecular Formula: C9H12BNO3
InChi Key: QJYYVSIRDJVQJW-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results