accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHClNOO

CAS RN 405910-34-1

IUPAC Name: 1-(4-chloro-2-nitrophenyl)piperazine
Synonyms: 1-(4-chloro-2-nitrophenyl)piperazine
Molecular Weight (g/mol): 241.68
Molecular Formula: C10H12ClN3O2
InChi Key: NZFQOXSALNTRDH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(Cl)=C1)N1CCNCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results