accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNNBr

CAS RN 40598-94-5

IUPAC Name: 3-bromo-2H-indazole
Synonyms: 3-bromo-2H-indazole
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: HTKXRTUKPXEALT-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=NN1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)