accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFBr

CAS RN 406-81-5

IUPAC Name: 4-bromo-1,1,1-trifluorobutane
Synonyms: 4-bromo-1,1,1-trifluorobutane
Molecular Weight (g/mol): 190.99
Molecular Formula: C4H6BrF3
InChi Key: DBCAQXHNJOFNGC-UHFFFAOYSA-N
SMILES: FC(F)(F)CCCBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)