accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClOCH3

CAS RN 40615-36-9

IUPAC Name: 1-[chloro(4-methoxyphenyl)phenylmethyl]-4-methoxybenzene
Synonyms: 1-[chloro(4-methoxyphenyl)phenylmethyl]-4-methoxybenzene
Molecular Weight (g/mol): 338.83
Molecular Formula: C21H19ClO2
InChi Key: JBWYRBLDOOOJEU-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
Molecular Weight (g/mol) 
  • (8)
  • (4)
Quantity 
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (6)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (2)
  • (6)
  • (8)
  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (6)
  • (2)
  • (3)