accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40635-66-3

IUPAC Name: 1-chloro-2-methyl-1-oxopropan-2-yl acetate
Synonyms: 1-chloro-2-methyl-1-oxopropan-2-yl acetate
Molecular Weight (g/mol): 164.59
Molecular Formula: C6H9ClO3
InChi Key: RBTCRFLJLUNCLL-UHFFFAOYSA-N
SMILES: CC(=O)OC(C)(C)C(Cl)=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)