CAS RN 40640-98-0
IUPAC Name:
ethyl 3-(4-bromophenyl)propanoate
Synonyms:
ethyl 3-(4-bromophenyl)propanoate
Molecular Weight (g/mol):
257.13
Molecular Formula:
C11H13BrO2
InChi Key:
STMHGPFYLQOGJD-UHFFFAOYSA-N
SMILES:
CCOC(=O)CCC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl 3-(4-bromophenyl)propionate, 95%
Ethyl 3-(4-bromophenyl)propionate, 95%
CAS: 40640-98-0 Molecular Formula: C11H13BrO2 Molecular Weight (g/mol): 257.13 MDL Number: MFCD07772961 InChI Key: STMHGPFYLQOGJD-UHFFFAOYSA-N PubChem CID: 12731927 IUPAC Name: ethyl 3-(4-bromophenyl)propanoate SMILES: CCOC(=O)CCC1=CC=C(Br)C=C1