accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONHNFFF

CAS RN 406476-31-1

IUPAC Name: 2-(4-carboxylatopiperidin-1-yl)-5-(trifluoromethyl)pyridin-1-ium
Synonyms: 2-(4-carboxylatopiperidin-1-yl)-5-(trifluoromethyl)pyridin-1-ium
Molecular Weight (g/mol): 274.24
Molecular Formula: C12H13F3N2O2
InChi Key: KNDSIDUPVUCATQ-UHFFFAOYSA-N
SMILES: [O-]C(=O)C1CCN(CC1)C1=CC=C(C=[NH+]1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results