CAS RN 40663-68-1
IUPAC Name:
4-(prop-2-en-1-yloxy)benzaldehyde
Synonyms:
4-(prop-2-en-1-yloxy)benzaldehyde
Molecular Weight (g/mol):
162.19
Molecular Formula:
C10H10O2
InChi Key:
TYNJQOJWNMZQFZ-UHFFFAOYSA-N
SMILES:
C=CCOC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Allyloxybenzaldehyde, 97%
4-Allyloxybenzaldehyde, 97%
CAS: 40663-68-1 Molecular Formula: C10H10O2 Molecular Weight (g/mol): 162.19 MDL Number: MFCD00014133 InChI Key: TYNJQOJWNMZQFZ-UHFFFAOYSA-N Synonym: 4-allyloxybenzaldehyde,4-allyloxy benzaldehyde,p-allyloxy benzaldehyde,p-allyloxybenzaldehyde,benzaldehyde, 4-2-propenyloxy,4-prop-2-en-1-yloxy benzaldehyde,4-prop-2-enyloxybenzaldehyde,4-allyloxybenzaldehyd,acmc-20ampt PubChem CID: 95942 IUPAC Name: 4-prop-2-enoxybenzaldehyde SMILES: C=CCOC1=CC=C(C=O)C=C1