accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 40665-68-7

IUPAC Name: dimethyl (2-oxo-3-phenoxypropyl)phosphonate
Synonyms: dimethyl 2-oxo-3-phenoxypropylphosphonate
Molecular Weight (g/mol): 258.21
Molecular Formula: C11H15O5P
InChi Key: NQTSTBMCCAVWOS-UHFFFAOYSA-N
SMILES: COP(=O)(CC(=O)COC1=CC=CC=C1)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results